missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (PIG45A2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61070
This item is not returnable.
View return policy
Description
IgM Monoclonal specifically detects IgM in Porcine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| IgM | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000 | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Mouse | |
| Unpurified | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunohistochemistry, Functional Assay | |
| PIG45A2 | |
| Unconjugated | |
| Ascites with 0.09% Sodium Azide | |
| IGHM | |
| Pig immunoglobulins | |
| 0.1 mg | |
| Primary | |
| Pig, Goat, Sheep | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction