missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (SPM557), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34422MFV450
This item is not returnable.
View return policy
Description
IgM Monoclonal antibody specifically detects IgM in Human samples. It is validated for Flow Cytometry
Specifications
| IgM | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| SPM557 | |
| Flow Cytometry | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Recombinant heavy chain of human IgM | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction