missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Antibody (V53P3H2*B2), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50439AF488
This item is not returnable.
View return policy
Description
IgM Monoclonal specifically detects IgM in Fish samples. It is validated for ELISA.
Specifications
| IgM | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IGHM | |
| Ovalbumin-conjugated synthetic peptide | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA | |
| V53P3H2*B2 | |
| ELISA | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Fish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction