missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF11 Monoclonal Antibody (OTI3A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803775
This item is not returnable.
View return policy
Description
Functions as a cell adhesion molecule through homophilic interaction. Stimulates cell growth.Specifications
| IGSF11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5DX21 | |
| IGSF11 | |
| Human recombit protein fragment corresponding to amino acids 23-241 of human IGSF11 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI3A4 | |
| Unconjugated | |
| IGSF11 | |
| BT-IgSF, CT119, CXADRL1, Igsf13, VSIG3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 152404 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?