missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF11 Mouse anti-Human, Clone: OTI2H1, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803797S
This item is not returnable.
View return policy
Description
IGSF11 Monoclonal antibody specifically detects IGSF11 in Human samples. It is validated for Western BlotSpecifications
| IGSF11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5DX21 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 152404 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2H1 | |
| Unconjugated | |
| Igsf11 | |
| 1700025L02Rik; brain and testis-specific immunoglobin superfamily protein; brain and testis-specific immunoglobulin superfamily protein; BTIGSF; Bt-IGSF; cancer/testis antigen 119; CT119; CXADR like 1; CXADRL1; Igsf11; Igsf13; Immunoglobulin superfamily member 11; immunoglobulin superfamily, member 11; immunoglobulin superfamily, member 13; V-set and immunoglobulin domain containing 3; V-set and immunoglobulin domain-containing protein 3; VSIG3 | |
| Human recombinant protein fragment corresponding to amino acids 23-241 of human IGSF11 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction