missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IGSF2/CD101 Antibody (8D4D3), Novus Biologicals™
Mouse Monoclonal Antibody
€ 287.00 - € 511.00
Specifications
| Antigen | IGSF2/CD101 |
|---|---|
| Clone | 8D4D3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30030479
|
Novus Biologicals™
NBP3-27139-0.025ml |
0.025 mL |
€ 287.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032757
|
Novus Biologicals™
NBP3-27139-0.1ml |
0.1 mL |
€ 511.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGSF2/CD101 Monoclonal antibody specifically detects IGSF2/CD101 in Human samples. It is validated for ELISA,Western BlotSpecifications
| IGSF2/CD101 | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, leukocyte surface protein, member 2 | |
| Purified recombinant fragment of human IGSF2/CD101 (AA: extra 22-168) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 8D4D3 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 9398 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title