missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF3 Antibody (503621), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4788C
This item is not returnable.
View return policy
Description
IGSF3 Monoclonal antibody specifically detects IGSF3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| IGSF3 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3321 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, CyTOF | |
| 503621 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| EWI-3, glu-Trp-Ile EWI motif-containing protein 3, immunoglobulin superfamily, member 3, member 3, V8 | |
| Mouse myeloma cell line NS0-derived recombinant human IGSF3, Gln20-Ala1125, Accession # AAI44133 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction