missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF4A/SynCAM1/CADM1 Antibody (41N3D3), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-14841AF488
This item is not returnable.
View return policy
Description
IGSF4A/SynCAM1/CADM1 Monoclonal specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for Western Blot.
Specifications
| IGSF4A/SynCAM1/CADM1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Q9BY67 | |
| CADM1 | |
| A portion of amino acids 150-200 of human CADM1 was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers, Neuroscience | |
| 23705 | |
| Human | |
| Purified |
| Western Blot | |
| 41N3D3 | |
| Western Blot | |
| BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in Human and Primates. 98% in Mouse, Dog, Rabbit, Horse and Chinese Hamster. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction