missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF4B/SynCAM3/CADM3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | IGSF4B/SynCAM3/CADM3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698372
|
Novus Biologicals
NBP2-98284-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618754
|
Novus Biologicals
NBP2-98284-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGSF4B/SynCAM3/CADM3 Polyclonal antibody specifically detects IGSF4B/SynCAM3/CADM3 in Mouse samples. It is validated for Western Blot, ELISASpecifications
| IGSF4B/SynCAM3/CADM3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 57863 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| BIgR, Brain immunoglobulin receptor, cell adhesion molecule 3, IgSF4B, IGSF4BSynCAM3, Immunoglobulin superfamily member 4B, member 4B, NECL-1, NECL1TSLC1-like protein 1, Nectin-like protein 1, Synaptic cell adhesion molecule 3, synCAM3, TSLC1-like 1, TSLL1SYNCAM3 | |
| Produced in rabbits immunized with purified, recombinant Mouse IGSF4B/SynCAM3/CADM3 (Accession#: NP_444429.1; Met1-His328) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title