missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IGSF8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552875

Product Code. 15780821

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PDVLQVSAAP PGPRGRQAPT SPPRMTVHEG QELALGCLAR TSTQKHTHLA VSFGRSVPEA PVGRSTLQEV VGIRSDLAVE AGAPYAERLA AGELRLGKEG TDRYRMVVGG AQAGDAGTYH CTAAEWIQDP DGSWAQIAEK RAVLAHVDVQ Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

This gene encodes a member the EWI subfamily of the immunoglobulin protein superfamily. Members of this family contain a single transmembrane domain, an EWI (Glu-Trp-Ile)-motif and a variable number of immunoglobulin domains. This protein interacts with the tetraspanins CD81 and CD9 and may regulate their role in certain cellular functions including cell migration and viral infection. The encoded protein may also function as a tumor suppressor by inhibiting the proliferation of certain cancers. Alternate splicing results in multiple transcript variants that encode the same protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IGSF8
Polyclonal
Unconjugated
IGSF8
AA033172; AU041109; CD316; CD81 partner 3; CD81P3; ESTM34; EWI2; EWI-2; Glu-Trp-Ile EWI motif-containing protein 2; IGSF8; Immunoglobulin superfamily member 8; immunoglobulin superfamily, member 8; Kct4; KCT-4; Keratinocyte-associated transmembrane protein 4; keratinocytes-associated transmembrane protein 4; LIR-D1; PG regulatory-like protein; PGRL; Prostaglandin regulatory-like protein; R75384
Rabbit
Antigen affinity chromatography
RUO
93185
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q969P0
IGSF8
Recombinant protein corresponding to Human IGSF8. Recombinant protein control fragment (Product #RP-89909).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.