missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IGSF9B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15557182
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15557182 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15557182 Supplier Invitrogen™ Supplier No. PA546478

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: QDENVYFQND LKLRVRILID GTLIIFRVKP EDSGKYTCVP SNSLGRSPSA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 91%.

IGSF9B is a protein coding gene. Transmembrane protein which is abundantly expressed in interneurons, where it may regulate inhibitory synapse development. May mediate homophilic cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF9B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IGSF9B
Gene Accession No. Q9UPX0
Gene Alias Gm508; IgSF9B; igsf9l; igsf9l protein; im:7148508; immunoglobulin superfamily member 9B; immunoglobulin superfamily, member 9b; immunoglobulin superfamily, member 9b; protein turtle homolog A; Kiaa1030; LOW QUALITY PROTEIN: protein turtle homolog B; mKIAA1030; Protein turtle homolog B; RGD1564717; si:dkey-49h9.6
Gene Symbols IGSF9B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human FGFRL1 (aa 266-315).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22997
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.