Learn More
IgY Mouse anti-Chicken, Unconjugated, Clone: 1E12G2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67405-1-IG-150UL
Product Code. 16820016
  • € 189.00 / 150µL
Description
The Chicken IgY antibody from Proteintech is a mouse monoclonal antibody generated with peptide of chicken Chicken IgY. This antibody recognizes chicken antigen. The Chicken IgY antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
IgY
Monoclonal
1000 μg/mL
Mouse
Primary
IgG2a
Western Blot, ELISA
1E12G2
Unconjugated
150 μL
Chicken