missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXFP3/RLN3R1/SALPR Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97628UV
This item is not returnable.
View return policy
Description
RXFP3/RLN3R1/SALPR Polyclonal specifically detects RXFP3/RLN3R1/SALPR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RXFP3/RLN3R1/SALPR | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| G protein-coupled receptor SALPR, GPCR135MGC141998, G-protein coupled receptor SALPR, relaxin 3 receptor 1, Relaxin family peptide receptor 3RLN3 receptor 1, relaxin/insulin-like family peptide receptor 3, relaxin-3 receptor 1, RLN3R1MGC142000, RXFPR3, SALPRG-protein coupled receptor GPCR135, somatostatin and angiotensin-like peptide receptor, Somatostatin- and angiotensin-like peptide receptor | |
| Produced in rabbits immunized with E. coli-derived Human RXFP3/RLN3R1/SALPR fragment. | |
| 0.1 mL | |
| GPCR | |
| 51289 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction