missing translation for 'onlineSavingsMsg'
Learn More

Proteintech IHCeasy Band 3/AE1 Ready-To-Use IHC Kit

Product Code. 17610267 Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17610267

Brand: Proteintech KHC020050T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-To-Use IHC Kit

KHC0200 is a ready-to-use IHC kit for staining of Band 3/AE1. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.

Specifications

Clonality Monoclonal
Target Species Human
Quantity 50 Tests
For Use With (Application) Immunohistochemistry (IHC)
Species Mouse
Content And Storage All the reagents are stored at 2°C to 8°C. The kit is stable for 6 months from the date of receipt.
Kit Contents
  • Antigen Retrieval Buffer(100 mL, 50 x)
  • Washing Buffer(100 mL x2, 20 x)
  • Blocking Buffer(5 mL, RTU)
  • Rabbit Polyclonal Primary Antibody(5 mL, RTU)
  • Polymer-HRP-Goat anti-Mouse Secondary Antibody(5 mL, RTU)
  • Chromogen Component A(0.2 mL, RTU)
  • Chromogen Component B(4 mL, RTU)
  • Signal Enhancer(5 mL, RTU)
  • Counter Staining Reagent(5 mL, RTU)
  • Mounting Media(5 mL, RTU)
  • Datasheet (1 Copy)
  • Manual(1 Copy)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.