Learn More
Proteintech IHCeasy CSNK2B Ready-To-Use IHC Kit
Product Code. 18028955 Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Product Code. 18028955
Brand: Proteintech KHC165050TS
KHC1650 is a ready-to-use IHC kit for staining of CSNK2B.
KHC1650 is a ready-to-use IHC kit for staining of CSNK2B. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Quantity 50 Tests
Form Liquid