missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ihfA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144577
This item is not returnable.
View return policy
Description
ihfA Polyclonal Antibody for Western Blot, ELISA
This protein is one of the two subunits of integration host factor, a specific DNA-binding protein that functions in genetic recombination as well as in transcriptional and translational control.
Specifications
| ihfA | |
| Polyclonal | |
| Unconjugated | |
| ihfA | |
| himA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 61280954 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.65 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P64393 | |
| ihfA | |
| Recombinant Neisseria meningitidis serogroup B ihfA protein (1-100aa). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction