Learn More
IkB-alpha Antibody (6A920), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56507MFV500
This item is not returnable.
View return policy
Description
IkB-alpha Monoclonal antibody specifically detects IkB-alpha in Human, Mouse, Rat samples. It is validated for Flow Cytometry