missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IkB-alpha, p Ser32 Antibody (SR1656), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21533-100ul
This item is not returnable.
View return policy
Description
IkB-alpha Monoclonal antibody specifically detects IkB-alpha in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| IkB-alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1656 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
| A synthesized peptide derived from human phospho-IKB alpha [Ser32] (Uniprot #: P25963) | |
| 100 μg | |
| Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction | |
| 4792 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur