missing translation for 'onlineSavingsMsg'
Learn More

IKBKAP Antibody - BSA Free, Novus Biologicals™

Product Code. 18042597 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18042597 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18042597 Supplier Novus Biologicals Supplier No. NBP2118010.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

MIS/AMH Monoclonal specifically detects MIS/AMH in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIS/AMH
Applications Western Blot
Classification Monoclonal
Clone RM0288-8D29
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. P27106.1
Gene Alias MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance
Gene Symbols AMH
Host Species Rat
Immunogen Mouse AMH
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.