missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKBKAP Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-214H
This item is not returnable.
View return policy
Description
IKBKAP Polyclonal antibody specifically detects IKBKAP in Human samples. It is validated for Western Blot
Specifications
| IKBKAP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| HRP | |
| DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1 | |
| C-terminus of the human IKAP protein (proprietary) | |
| 0.1 mL | |
| Cancer, Transcription Factors and Regulators | |
| 8518 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction