missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKBKAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 528.15
Specifications
| Antigen | IKBKAP |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664396
|
Novus Biologicals
NBP2-48973-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649117
|
Novus Biologicals
NBP2-48973 |
0.1 mL |
€ 559.00 € 528.15 / 0.10mL Save € 30.85 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IKBKAP Polyclonal antibody specifically detects IKBKAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| IKBKAP | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS (pH 7.2), 40% Glycerol | |
| 8518 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RLHVLCQGWHYLAYDWHWTTDRSVGDNSSDLSNVAVIDGNRVLVTVFRQTVVPPPMCTYQLLFPHPVNQVTFLAHPQKSNDLAVLDASNQISVYKCGDCPSADPTVKLGAVG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title