Learn More
IKBKG Mouse anti-Human, Clone: OTI2F2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812458
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IKK gamma is the regulatory subunit of the inhibitor of the kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.Specifications
| IKBKG | |
| Monoclonal | |
| Unconjugated | |
| IKBKG | |
| AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8517 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2F2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9Y6K9 | |
| IKBKG | |
| Full length human recombit protein of human IKBKG produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.