Learn More
Invitrogen™ IKK alpha Recombinant Rabbit Monoclonal Antibody (23GB765), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553362
 View more versions of this product
Product Code. 30089558
  • € 590.00 / 100µL
Description
CHUK (IKK alpha) is a serine/threonine kinase involved in signal transduction and cellular communication. It belongs to the cytokine activated protein complex that is an inhibitor of the NF-kappa B complex. IKK alpha along with IKK beta and a regulatory protein NEMO (IKK gamma) phosphorylates IkB proteins which targets the later for ubiquitin mediated degradation. It is involved with several cellular processes with extensive studies on its role in inflammation and cell survival.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IKK alpha
Recombinant Monoclonal
Unconjugated
CHUK
AI256658; CHUK; Chuk1; component of inhibitor of nuclear factor kappa B kinase complex; conserved helix-loop-helix ubiquitous kinase; EC 2.7.11.10; Fbx24; Fbxo24; FLJ40509; I kappa B kinase 1; i kappa-B kinase alpha; ikappaB kinase; I-kappa-B kinase 1; I-kappa-B kinase 2; IkappaB kinase alpha; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; IkB kinase alpha subunit; IkB kinase-alpha; ikBKA; IKBKB; IKK alpha; IKK1; IKK2; Ikka; IKK-A; IKK-a kinase; IKK-alpha; IKKB; IKK-B; IKK-beta; Inhibitor of nuclear factor kappa-B kinase subunit alpha; kinase beta; MGC131801; NFKBIKA; NFKBIKB; Nuclear factor NFkappaB inhibitor kinase alpha; Nuclear factor NF-kappa-B inhibitor kinase alpha; TCF16; TCF-16; Transcription factor 16
Rabbit
Affinity chromatography
RUO
1147, 12675, 309361
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB765
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O15111, Q60680
CHUK
A synthetic peptide of human IKK alpha (1-59AA).
100 μL
Primary
Human, Mouse, Rat