missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK beta Antibody (10A9B6), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56513AF488
This item is not returnable.
View return policy
Description
IKK beta Monoclonal specifically detects IKK beta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
Specifications
| IKK beta | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| An approx. 87 kDa band should be observed. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 10A9B6 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular) | |
| O14920 | |
| IKBKB | |
| Full-length human IKKb/IKK2 recombinant protein was used as immunogen (NP_001547). | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity, mTOR Pathway, Phospho Specific, Protein Kinase | |
| 3551 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction