missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK beta Antibody (10AG2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56509CL1
This item is not returnable.
View return policy
Description
IKK beta Monoclonal antibody specifically detects IKK beta in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunoblot,Simple Western,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| IKK beta | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta | |
| Full-length recombinant human IKK beta protein (NP_001547). | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity, mTOR Pathway, Phospho Specific, Protein Kinase | |
| 3551 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 10AG2 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction