missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK epsilon/IKBKE Antibody (OTI3C5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71490G
This item is not returnable.
View return policy
Description
IKK epsilon/IKBKE Monoclonal specifically detects IKK epsilon/IKBKE in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| IKK epsilon/IKBKE | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IKBKE | |
| Recombinant protein fragment corresponding to amino acids 474-716 of human IKK epsilon/IKBKE (NP_054721) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3C5 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase epsilon, IkBKE, IKK-E, IKKEMGC125297, IKK-epsilon, IKKI, IKK-iMGC125295, Inducible I kappa-B kinase, inducible IkappaB kinase, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon, inhibitor of nuclear factor kappa-B kinase subunit epsilon, KIAA0151IKK-related kinase epsilon, MGC125294 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Phospho Specific, Protein Kinase | |
| 9641.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction