missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK gamma Antibody (46B844), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56532G
This item is not returnable.
View return policy
Description
IKK gamma Monoclonal specifically detects IKK gamma in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| IKK gamma | |
| Monoclonal | |
| DyLight 488 | |
| NP_001093326 | |
| IKBKG | |
| Full-length human IKK gamma protein. | |
| 0.1 mL | |
| Apoptosis, Cancer, Immunology, Innate Immunity | |
| 8517 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 46B844 | |
| Western Blot, Flow Cytometry | |
| FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| A 48 kDa band is observed. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction