missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK gamma Antibody (46B844), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33220JF549
This item is not returnable.
View return policy
Description
IKK gamma Monoclonal specifically detects IKK gamma in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| IKK gamma | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IKBKG | |
| Full-length human IKK gamma protein. | |
| 0.1 mL | |
| Primary | |
| A 48 kDa band is observed. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| 46B844 | |
| Western Blot, Flow Cytometry | |
| FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator | |
| Mouse | |
| Protein G purified | |
| Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific | |
| 8517.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction