Learn More
IKZF3 Mouse anti-Human, Clone: OTI2B9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804393
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
IKZF3 is a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.Specifications
| IKZF3 | |
| Monoclonal | |
| Unconjugated | |
| IKZF3 | |
| AIO, AIOLOS, ZNFN1A3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 22806 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UKT9 | |
| IKZF3 | |
| Human recombit protein fragment corresponding to amino acids 1-301 of human IKZF3 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.