Learn More
Invitrogen™ IKZF3 Recombinant Rabbit Monoclonal Antibody (4U7F9)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542889
Product Code. 17866322
  • € 502.00 / 100µL
Description
Positive test controls include: Raji, Mouse spleen, Mouse thymus, Rat lung. Immunogen sequence: IYQQNHMVLS RARNGMPLLK EVPRSYELLK PPPICPRDSV KVINKEGEVM DVYRCDHCRV LFLDYVMFTI HMGCHGFRDP FECNMCGYRS HDRYEFSSHI ARGEHRALLK.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IKZF3
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O08900, Q9UKT9
Ikzf3
Recombinant fusion protein containing a sequence corresponding to amino acids 400-509 of human IKZF3 (Q9UKT9).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
4U7F9
Unconjugated
Ikzf3
5830411O07Rik; AIO; AIOLOS; IKAROS family zinc finger 3; IKAROS family zinc finger 3 (Aiolos); ikaros family zinc finger protein 3; Ikzf3; Zfpn1a3; zinc finger DNA binding protein Aiolos; zinc finger protein Aiolos; zinc finger protein, subfamily 1A, 3 (Aiolos); Znfn1a3
Rabbit
Affinity chromatography
RUO
22780, 22806, 303511
-20°C, Avoid Freeze/Thaw Cycles