missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IKZF3 Recombinant Rabbit Monoclonal Antibody (JE62-81)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544810
This item is not returnable.
View return policy
Description
IKZF3 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.
Specifications
| IKZF3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9UKT9 | |
| Ikzf3 | |
| Recombinant protein within human IKZF3 aa 360-509/509. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| JE62-81 | |
| Unconjugated | |
| Ikzf3 | |
| 5830411O07Rik; AIO; AIOLOS; IKAROS family zinc finger 3; IKAROS family zinc finger 3 (Aiolos); ikaros family zinc finger protein 3; Ikzf3; Zfpn1a3; zinc finger DNA binding protein Aiolos; zinc finger protein Aiolos; zinc finger protein, subfamily 1A, 3 (Aiolos); Znfn1a3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22806 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction