Learn More
Invitrogen™ IKZF3 Recombinant Rabbit Monoclonal Antibody (4U7F9)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542889
Product Code. 17866322
  • € 502.00 / 100µL
Description
Positive test controls include: Raji, Mouse spleen, Mouse thymus, Rat lung. Immunogen sequence: IYQQNHMVLS RARNGMPLLK EVPRSYELLK PPPICPRDSV KVINKEGEVM DVYRCDHCRV LFLDYVMFTI HMGCHGFRDP FECNMCGYRS HDRYEFSSHI ARGEHRALLK.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
IKZF3
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O08900, Q9UKT9
Ikzf3
Recombinant fusion protein containing a sequence corresponding to amino acids 400-509 of human IKZF3 (Q9UKT9).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
4U7F9
Unconjugated
Ikzf3
5830411O07Rik; AIO; AIOLOS; IKAROS family zinc finger 3; IKAROS family zinc finger 3 (Aiolos); ikaros family zinc finger protein 3; Ikzf3; Zfpn1a3; zinc finger DNA binding protein Aiolos; zinc finger protein Aiolos; zinc finger protein, subfamily 1A, 3 (Aiolos); Znfn1a3
Rabbit
Affinity chromatography
RUO
22780, 22806, 303511
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Product Certifications
Promotions
Promotions
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis