missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IKZF4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15998174
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15998174 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15998174 Supplier Invitrogen™ Supplier No. PA565433

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VNSGGYEKDVE LVAHHSLEPG FGSSLAFVGA EHLRPLRLPP TNCISELTPV ISSVYTQMQP LPGRLELPGS REAGEGPEDL ADGGPLLYRP RGPLTDPGA Highest antigen sequence identity to the following orthologs - mouse 90%, rat 91%.

DNA-binding protein that binds to the 5'GGGAATRCC-3' Ikaros-binding sequence. Transcriptional repressor. Interacts with SPI1 and MITF to repress transcription of the CTSK and ACP5 promoters via recruitment of corepressors SIN3A and CTBP2. May be involved in the development of central and peripheral nervous systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKZF4
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IKZF4
Gene Accession No. Q9H2S9
Gene Alias A630026H08Rik; Eos; IKAROS family zinc finger 4; IKAROS family zinc finger 4 (Eos); ikaros family zinc finger protein 4; Ikzf4; KIAA1782; Zfpn1a4; Zinc finger protein Eos; zinc finger protein, subfamily 1A, 4; zinc finger protein, subfamily 1A, 4 (Eos); zinc finger transcription factor Eos; ZNFN1A4
Gene Symbols IKZF4
Host Species Rabbit
Immunogen Recombinant Human IKZF4. Recombinant protein control fragment (Product #RP-104817).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64375
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.