missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 alpha/IL-1F1 Antibody (3405) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2001JF585
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Monoclonal antibody specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Flow Cytometry,Neutralization,CyTOF
Specifications
| IL-1 alpha/IL-1F1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| E. coli-derived recombinant human IL-1 alpha /IL-1F1, Ser113-Ala271, Accession # P01583 | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| 3552 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Neutralization, CyTOF | |
| 3405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction