missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 beta/IL-1F2 Antibody (1027B) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8406JF585
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Monoclonal antibody specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
| IL-1 beta/IL-1F2 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| E.coli-derived recombinant human IL-1β/IL-1F2, Ala117-Ser269, Accession # P01584 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry (Intracellular Staining) | |
| 1027B | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cardiovascular Biology, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Lipid and Metabolism, Metastasis | |
| 3553 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction