missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 RI Antibody (129304), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7712L
This item is not returnable.
View return policy
Description
IL-1 RI Monoclonal antibody specifically detects IL-1 RI in Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
| IL-1 RI | |
| Monoclonal | |
| DyLight 550 | |
| CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-1 RI, Leu20-Lys338, Accession # P13504 | |
| 0.5 mL | |
| Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
| 3554 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry | |
| 129304 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction