Learn More
IL-1 RI Antibody (129304), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB7712PEATT594
Product Code. 30036094
  • ONLINE EXCLUSIVE PRICE
    € 1220.00
    € 1150.80 / 0.50mL
    Save € 69.20
    5% Off
Description
IL-1 RI Monoclonal antibody specifically detects IL-1 RI in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-1 RI
Monoclonal
PE/ATTO 594
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry
129304
Flow Cytometry
CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Mouse myeloma cell line NS0-derived recombinant mouse IL-1 RI, Leu20-Lys338, Accession # P13504
0.5 mL
Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific
3554
Store at 4C in the dark. Do not freeze.
IgG2b