missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 RII Antibody (20), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89273AF488
This item is not returnable.
View return policy
Description
IL-1 RII Monoclonal antibody specifically detects IL-1 RII in Human samples. It is validated for ELISA
Specifications
| IL-1 RII | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 20 | |
| ELISA | |
| beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-1 RII (Met 1-Glu 343; NP_004624.1). | |
| 0.1 mL | |
| Cancer, Phospho Specific, Signal Transduction | |
| 7850 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction