missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 RII Antibody (RM0014-7D7), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-85472DL594
This item is not returnable.
View return policy
Description
IL-1 RII Monoclonal antibody specifically detects IL-1 RII in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| IL-1 RII | |
| Monoclonal | |
| DyLight 594 | |
| beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725 | |
| Mouse IL-1 RII (extracellular domain) | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 7850 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0014-7D7 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction