missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Antibody (2050B), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9210R
This item is not returnable.
View return policy
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human samples. It is validated for Neutralization, Flow Cytometry
Specifications
| IL-10 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Neutralization, Flow Cytometry | |
| 2050B | |
| Neutralization, Intracellular Staining by Flow Cytometry | |
| CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10, Met1-Asn178, Accession # P22301 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3586 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction