Learn More
IL-10 Antibody (BT-063) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28584JF585
Product Code. 30169384
  • € 431.00 / 0.10mL
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-10
Monoclonal
Janelia Fluor 585
CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
IL-10
0.1 mL
Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity
3586
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
BT-063
50mM Sodium Borate
Human
Protein A purified