missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Antibody (CC320), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63026UV
This item is not returnable.
View return policy
Description
IL-10 Monoclonal specifically detects IL-10 in Rat, Bovine, Equine, Goat, Sheep samples. It is validated for Flow Cytometry, ELISA.
Specifications
| IL-10 | |
| Monoclonal | |
| DyLight 350 | |
| CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Clone CC320 has been shown to neutralise the activity of bovine IL-10 as measured by the inhibition of the inhibitory activity of IL-10 on IFN gamma synthesis (2). | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA | |
| CC320 | |
| Flow Cytometry, ELISA | |
| IL10 | |
| Plasmid cDNA encoding bovine IL-10 | |
| 0.25 mL | |
| Adaptive Immunity, Apoptosis, Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3586 | |
| Bovine, Equine, Goat, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction