missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Antibody (JES3-9D7), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-77379APC
This item is not returnable.
View return policy
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human, Cynomolgus Monkey, Macaque, Virus samples. It is validated for Western Blot, ELISA, ELISA Capture (Matched Antibody Pair)
Specifications
| IL-10 | |
| Monoclonal | |
| APC | |
| CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| COS-expressed human IL-10 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3586 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, ELISA Capture (Matched Antibody Pair) | |
| JES3-9D7 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey, Macaque, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction