missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 R beta Antibody (009), Alexa Fluor™ 700 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90735AF700
This item is not returnable.
View return policy
Description
IL-10 R beta Monoclonal antibody specifically detects IL-10 R beta in Mouse samples. It is validated for ELISA, ELISA
Specifications
| IL-10 R beta | |
| Monoclonal | |
| Alexa Fluor 700 | |
| CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-10 R beta (NP_032375.2; Met1-Ser222). | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 3588 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 009 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido