missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-10 Recombinant Rabbit Monoclonal Antibody (6U0G7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542656
This item is not returnable.
View return policy
Description
Positive test controls include: THP-1, Mouse spleen, Rat thymus. Immunogen sequence: MHSSALLCCL VLLTGVRASP GQGTQSENSC THFPGNLPNM LRDLRDAFSR VKTFFQMKDQ LDNLLLKESL LEDFKGYLGC QALSEMIQFY LEEVMPQAEN.
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
Specifications
| IL-10 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22301, P29456 | |
| IL10 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Rat, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 6U0G7 | |
| Unconjugated | |
| IL10 | |
| CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25325, 3586 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction