missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10R alpha Antibody (1B1.3a), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-63577PE
This item is not returnable.
View return policy
Description
IL-10R alpha Monoclonal specifically detects IL-10R alpha in Mouse, Human (Negative) samples. It is validated for Flow Cytometry.
Specifications
| IL-10R alpha | |
| Monoclonal | |
| PE | |
| CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-63577 recognizes mouse CDw210a, a cell surface glycoprotein of approximately 110KD, also known as Interleukin-10 receptor 1 (IL-10R1). The epitope recognised by this antibody has been localised to the ligand binding domain within the extracellular region of mouse CDw210a. Mouse CDw210a is expressed by a range of cells including thymocytes, T cells, B cells and monocytes. This clone is reported to neutralize the effects of IL-10 (1). | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 1B1.3a | |
| Flow Cytometry | |
| IL10RA | |
| Purified recombinant extracellular region of mouse CD210 | |
| 0.1 mL | |
| Cytokine Research | |
| 3587 | |
| Mouse, Human (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction