Learn More
IL-10R alpha Antibody (714212), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB2742P
Product Code. 18696503
  • € 670.00 / 0.10mL
Description
IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-10R alpha
Monoclonal
PE
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
714212
Western Blot, Flow Cytometry, CyTOF-ready
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Mouse myeloma cell line NS0-derived and S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10 R alpha , His22-Asn235, Accession # Q13651
0.1 mL
Cytokine Research
3587
Store at 4°C in the dark.
IgG2b