Learn More
IL-10R alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-21324-100ul
 View more versions of this product
Product Code. 18654474
  • € 672.00 / 100µL
Description
IL-10R alpha Polyclonal antibody specifically detects IL-10R alpha in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-10R alpha
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
This antibody was developed against Recombinant Protein corresponding to amino acids: HGTELPSPPSVWFEAEFFHHILHWTPIPNQSESTCYEVALLRYGIESWNSISNCSQTLSYDL
100 μg
Cytokine Research
3587
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol