missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-11 Antibody (22626) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB218JF585
This item is not returnable.
View return policy
Description
IL-11 Monoclonal antibody specifically detects IL-11 in Human samples. It is validated for Western Blot,Neutralization
Specifications
| IL-11 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11Oprelvekin, interleukin 11, interleukin-11 | |
| E. coli-derived recombinant human IL-11, Pro22-Leu199, Accession # P20809 | |
| 0.5 mL | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| 3589 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Neutralization | |
| 22626 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction