missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-11 Antibody (MM0361-9T27), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11655FR
This item is not returnable.
View return policy
Description
IL-11 Monoclonal specifically detects IL-11 in Human samples. It is validated for Western Blot.
Specifications
| IL-11 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IL11 | |
| Human recombinant IL-11 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0361-9T27 | |
| Western Blot | |
| Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11Oprelvekin, interleukin 11, interleukin-11 | |
| Mouse | |
| Protein G purified | |
| Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity | |
| 3589 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction